2-碘乙醇 99% stabilised

供应商: Thermo Fisher Scientific
Danger

175500100 175500500
ACRO175500100EA 0 CNY
ACRO175500100 ACRO175500500
2-碘乙醇 99% stabilised
2-碘乙醇
分子式: C₂H₅IO
分子量: 171.97 g/mol
沸点: 172…175 °C (760 mmHg)
密度: 2.205 g/cm³
闪点: 65 °C
存储条件: Refrigerator
MDL编号: MFCD00002830
CAS号: 624-76-0
EINECS: 210-861-6
UN号: 2810
ADR: 6.1 II

现在订购

产品技术规格

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5690 to 1.5720 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR