乙酰氯 1 M in dichloromethane, AcroSeal®
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Acetyl chloride 1M in Dichloromethane
387741000
387748000
ACRO387741000EA
0
CNY
ACRO387741000
ACRO387748000
乙酰氯 1 M in dichloromethane, AcroSeal®
乙酰氯
分子式:
C₂H₃ClO 分子量: 78.5 g/mol 沸点: 50.9 溶点: -112 闪点: 5 存储条件: Ambient |
MDL编号:
MFCD00000719 CAS号: 75-36-5 EINECS: 200-865-6 UN号: 2922 ADR: 8 II Merck Index: 13,00087 |
产品技术规格
Appearance | Clear colorless Solution |
Titration Argentometric | 0.9 to 1.1 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...