三苯基膦氢溴酸盐 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 三苯基溴化膦

269090250 269091000 269090050
ACRO269090250EA 0 CNY
ACRO269090250 ACRO269091000 ACRO269090050
三苯基膦氢溴酸盐 99%
三苯基膦氢溴酸盐
分子式: C₁₈H₁₆BrP
分子量: 343.2 g/mol
溶点: 196…199 °C
存储条件: Ambient
MDL编号: MFCD00035107
CAS号: 6399-81-1
EINECS: 229-012-6
UN号: 3261
ADR: 8 III

现在订购

产品技术规格

Appearance White to light beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR