tert-Amyl ethyl ether ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
1,1-Dimethylpropyl ethyl ether, Ethyl tert-pentyl ether
A1673-25G
A1673-5G
TCIAA1673-25GEA
0
CNY
TCIAA1673-25G
TCIAA1673-5G
tert-Amyl ethyl ether ≥98.0% (by GC)
tert-Amyl ethyl ether
分子式:
C₇H₁₆O 沸点: 102°C |
MDL编号:
MFCD01727266 CAS号: 919-94-8 UN号: 3271 ADR: 3,II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...