3,5-二甲基吡啶 98+%

供应商: Thermo Fisher Scientific
Danger

125211000 125215000
ACRO125211000EA 0 CNY
ACRO125211000 ACRO125215000
3,5-二甲基吡啶 98+%
3,5-二甲基吡啶
分子式: C₇H₉N
沸点: 172,7 °C (1013 hPa)
溶点: –9 °C
密度: 0,93 g/cm³
闪点: 53 °C
存储条件: Ambient
MDL编号: MFCD00006404
CAS号: 591-22-0
EINECS: 209-708-6
UN号: 2920
ADR: 8,II

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5040 to 1.5060 (20°C, 589 nm)
GC ≥98.0 %
Sulfate (SO4) ≤0.05 %
Iron (Fe) ≤0.005 %
Acidity (HCl) ≤0.025 %
Insoluble matter ≤0.025 % (in water and ppt by NH4OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR