2,6-二叔丁基吡啶 97%

供应商: Thermo Fisher Scientific
Warning

204640050 204640250
ACRO204640050EA 0 CNY
ACRO204640050 ACRO204640250
2,6-二叔丁基吡啶 97%
2,6-二叔丁基吡啶
分子式: C₁₃H₂₁N
分子量: 191.32 g/mol
沸点: 209
溶点: 2…4 °C
密度: 0.852 g/cm³
闪点: 72 °C
存储条件: Refrigerator
MDL编号: MFCD00006306
CAS号: 585-48-8
EINECS: 209-557-6
Merck Index: 12,03086

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4740 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR