2,4-二甲基吡啶 98+%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2,4-卢剔啶
125181000
125185000
ACRO125181000EA
0
CNY
ACRO125181000
ACRO125185000
2,4-二甲基吡啶 98+%
2,4-二甲基吡啶
分子式:
C₇H₉N 分子量: 107.16 g/mol 沸点: 159 溶点: –60 °C 密度: 0.92 g/cm³ 闪点: 47 °C 存储条件: Ambient |
MDL编号:
MFCD00006337 CAS号: 108-47-4 EINECS: 203-586-8 UN号: 1992 ADR: 3 III |
产品技术规格
Appearance | Clear colorless to light yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4990 to 1.5010 (20°C, 589 nm) |
GC | ≥98.0 % |
Total volume capacity | ≥1.10 eq/l (OH) as is |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...