3-皮考林 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
3-甲基吡啶
131130010
131132500
131135000
ACRO131130010EA
0
CNY
ACRO131130010
ACRO131132500
ACRO131135000
3-皮考林 99%
3-皮考林
分子式:
C₆H₇N 分子量: 93.13 g/mol 沸点: 143…144 °C (760 mmHg) 溶点: –19 °C 密度: 0.95 g/cm³ 闪点: 38 °C 存储条件: Ambient |
MDL编号:
MFCD00006402 CAS号: 108-99-6 EINECS: 203-636-9 UN号: 2313 ADR: 3 III Merck Index: 13,07485 |
产品技术规格
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5050 to 1.5070 (20°C, 589 nm) |
GC | ≥98.5 % |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...