2-甲氧基吡啶 98%

供应商: Thermo Fisher Scientific
Warning

126100250 126101000 126100050
ACRO126100250EA 0 CNY
ACRO126100250 ACRO126101000 ACRO126100050
2-甲氧基吡啶 98%
2-甲氧基吡啶
分子式: C₆H₇NO
分子量: 109.13 g/mol
沸点: 142
密度: 1.04 g/cm³
闪点: 32 °C
存储条件: Ambient
MDL编号: MFCD00006262
CAS号: 1628-89-3
EINECS: 216-623-8
UN号: 1993
ADR: 3 III

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5050 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR