2-氟吡啶 98%

供应商: Thermo Fisher Scientific
Warning

119630500 119630050
ACRO119630500EA 0 CNY
ACRO119630500 ACRO119630050
2-氟吡啶 98%
2-氟吡啶
分子式: C₅H₄FN
分子量: 97.09 g/mol
沸点: 126 °C (753 mmHg)
密度: 1.12 g/cm³
闪点: 24 °C
存储条件: Ambient
MDL编号: MFCD00006224
CAS号: 372-48-5
EINECS: 206-757-5
UN号: 1993
ADR: 3 III

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4690 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR