2,3-二氯吡啶 99%

供应商: Thermo Fisher Scientific
Warning

113720100 113720500
ACRO113720100EA 0 CNY
ACRO113720100 ACRO113720500
2,3-二氯吡啶 99%
2,3-二氯吡啶
分子式: C₅H₃Cl₂N
沸点: 202,5…203.5 °C (1013 hPa)
溶点: 67…70 °C
存储条件: Ambient
MDL编号: MFCD00006229
CAS号: 2402-77-9
EINECS: 219-281-8
UN号: 2811
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥98.5 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR