氟化锆(IV) 99.5%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Zirconium (IV) fluoride
299790250
299791000
ACRO299790250EA
0
CNY
ACRO299790250
ACRO299791000
氟化锆(IV) 99.5%
氟化锆(IV)
分子式:
F₄Zr 分子量: 167.22 g/mol 沸点: 905 °C 溶点: 640 °C |
MDL编号:
MFCD00011307 CAS号: 7783-64-4 EINECS: 232-018-1 UN号: 3260 ADR: 8 II Merck Index: 12,10308 |
产品技术规格
Appearance | White Crystalline powder |
Assay | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...