二苯基二硒醚 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 二苯联硒

177570050 177570250
ACRO177570050EA 0 CNY
ACRO177570050 ACRO177570250
二苯基二硒醚 99%
二苯基二硒醚
分子式: C₁₂H₁₀Se₂
分子量: 312.13 g/mol
沸点: 377
溶点: 60…63 °C
存储条件: Ambient
MDL编号: MFCD00003001
CAS号: 1666-13-3
EINECS: 216-780-2
UN号: 3283
ADR: 6.1 II

现在订购

产品技术规格

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 63 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR