三溴化磷 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
溴化磷(III)
169470050
169475000
ACRO169470050EA
0
CNY
ACRO169470050
ACRO169475000
三溴化磷 99%
三溴化磷
分子式:
PBr₃ 分子量: 270.69 g/mol 沸点: 175 °C (760 mmHg) 溶点: –40 °C 密度: 2.88 g/cm³ 存储条件: Ambient |
MDL编号:
MFCD00011436 CAS号: 7789-60-8 EINECS: 232-178-2 UN号: 1808 ADR: 8 II Merck Index: 12,07514 |
产品技术规格
Appearance | Clear to slightly hazy colorless to yellow to light brown Liquid |
Titration Argentometric | ≥98.5 % (ex Br) |
Melting point | 110 to 116 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...