Tin(IV) chloride, anhydrous 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: Stannic chloride, Tin tetrachloride

197510010 197510100 197511000
ACRO197510010EA 0 CNY
ACRO197510010 ACRO197510100 ACRO197511000
Tin(IV) chloride, anhydrous 99%
Tin(IV) chloride
分子式: Cl₄Sn
分子量: 260.52 g/mol
沸点: 114 °C (760 mmHg)
溶点: –33 °C
密度: 2.22 g/cm³
存储条件: Ambient
MDL编号: MFCD00011242
CAS号: 7646-78-8
EINECS: 231-588-9
UN号: 1827
ADR: 8 II
Merck Index: 12,08929

现在订购

产品技术规格

Appearance Colorless to light yellow Liquid (May develop some turbidity or precipitate)
Titration Argentometric 98.5 to 101.5 %
Free chlorine ≤10 ppm
Arsenic (As) ≤40 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR