硫酸铊(I) 99.9+% (trace metals basis)

供应商: Thermo Fisher Scientific
Danger

Synonyms: Thallic sulphate, Dithallium sulphate

196940100 196940500
ACRO196940100EA 0 CNY
ACRO196940100 ACRO196940500
硫酸铊(I) 99.9+% (trace metals basis)
硫酸铊(I)
分子式: O₄STl₂
分子量: 504.83 g/mol
溶点: 632 °C
密度: 6.77 g/cm³
存储条件: Ambient
MDL编号: MFCD00011278
CAS号: 7446-18-6
EINECS: 231-201-3
UN号: 1707
ADR: 6.1 II
Merck Index: 12,09405

现在订购

产品技术规格

Appearance White to off-white Powder or crystals
Assay 80.0 to 84.0 % (Tl)
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR