3,5-二氯吡啶-4-甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 3,5-二氯异烟酸

D2600-1G D2600-5G
TCIAD2600-1GEA 0 CNY
TCIAD2600-1G TCIAD2600-5G
3,5-二氯吡啶-4-甲酸 ≥98.0% (by titrimetric analysis)
3,5-二氯吡啶-4-甲酸
分子式: C₆H₃Cl₂NO₂
MDL编号: MFCD01861975
CAS号: 13958-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04d10e59e774a8a5a8bb8bcc4b4bdf4b9e2bad7d&vid=04d10e59e774a8a5a8bb8bcc4b4bdf4b9e2bad7d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol (50vol%) very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR