4-乙基苯乙炔 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(4-Ethylphenyl)acetylene, 1-Ethyl-4-ethynylbenzene, 1-Ethynyl-4-ethylbenzene, (4-Ethylphenyl)ethyne
E0749-25G
E0749-5G
TCIAE0749-25GEA
0
CNY
TCIAE0749-25G
TCIAE0749-5G
4-乙基苯乙炔 ≥97.0% (by GC)
4-乙基苯乙炔
分子式:
C₁₀H₁₀ 沸点: 192°C 闪点: 71°C 存储条件: Refrigerator |
MDL编号:
MFCD00173887 CAS号: 40307-11-7 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...