Acetamidine hydrochloride 98+%

供应商: Thermo Fisher Scientific
Warning

170210100 170211000
ACRO170210100EA 0 CNY
ACRO170210100 ACRO170211000
Acetamidine hydrochloride 98+%
Acetamidine hydrochloride
分子式: C₂H₇ClN₂
分子量: 94.54 g/mol
溶点: 160…170 °C
存储条件: Ambient
MDL编号: MFCD00013016
CAS号: 124-42-5
EINECS: 204-700-9

现在订购

产品技术规格

Appearance White to cream Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric 98.0 to 102.0 %
HPLC 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR