1,8-二氮杂双环[5.4.0]十一碳-7-烯 98+%

供应商: Thermo Fisher Scientific
Danger

160610250 160611000 160615000
ACRO160610250EA 0 CNY
ACRO160610250 ACRO160611000 ACRO160615000
1,8-二氮杂双环[5.4.0]十一碳-7-烯 98+%
1,8-二氮杂双环[5.4.0]十一碳-7-烯
分子式: C₉H₁₆N₂
沸点: 261 °C (760 mmHg)
溶点: –70 °C
密度: 1,019 g/cm³
闪点: 116 °C
存储条件: Ambient
MDL编号: MFCD00006930
CAS号: 6674-22-2
EINECS: 229-713-7
UN号: 2922
ADR: 8,II

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5230 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR