靛红酸酐 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 衣托酸酐, N-羧鄰胺苯甲酐

122481000 122485000
ACRO122481000EA 0 CNY
ACRO122481000 ACRO122485000
靛红酸酐 98%
靛红酸酐
分子式: C₈H₅NO₃
溶点: 243 °C (dec.)
闪点: 308 °C
存储条件: Ambient
MDL编号: MFCD00006700
CAS号: 118-48-9
EINECS: 204-255-0

现在订购

产品技术规格

Appearance Beige to brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR