乙醛酸一水合物 98%, pure

供应商: Thermo Fisher Scientific
Danger

Synonyms: 甲酰甲酸, 氧代乙酸

120170250 120171000 120175000
ACRO120170250EA 0 CNY
ACRO120170250 ACRO120171000 ACRO120175000
乙醛酸一水合物 98%, pure
乙醛酸一水合物
分子式: C₂H₂O₃·1H₂O
分子量: 92.05 g/mol
溶点: 48…52 °C
闪点: >110 °C
存储条件: Ambient
MDL编号: MFCD00127974
CAS号: 563-96-2
EINECS: 206-058-5
UN号: 3261
ADR: 8 II

现在订购

产品技术规格

Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 48 to 52 °C
Titration with NaOH ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR