Sodium tetrachloropalladate(II) (∼36% Pd)
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Palladium sodium chloride, Sodium palladium (II) chloride, Disodium tetrachloropalladate
195130050
195130250
195130010
ACRO195130050EA
0
CNY
ACRO195130050
ACRO195130250
ACRO195130010
Sodium tetrachloropalladate(II) (∼36% Pd)
Sodium tetrachloropalladate(II)
分子式:
Cl₄Na₂Pd 分子量: 294.21 g/mol 存储条件: Ambient |
MDL编号:
MFCD00003487 CAS号: 13820-53-6 EINECS: 237-502-6 UN号: 1759 ADR: 8 III |
产品技术规格
Appearance | Green-brown to brown Powder or granules and/or chunks |
Assay | 35 to 37 % (Pd) |
Total trace metal impurities | ≤300 ppm (Pt + Ru + Rh + Ir) |
Calcium (Ca) | ≤50 ppm |
Copper (Cu) | ≤50 ppm |
Gold (Au) | ≤100 ppm |
Iron (Fe) | ≤50 ppm |
Magnesium (Mg) | ≤50 ppm |
Silicon (Si) | ≤50 ppm |
Silver (Ag) | ≤100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...