过氧化钠 ≥93.0%, powder, AnalaR NORMAPUR® analytical reagent
供应商: VWR Chemicals
|
Danger
|
Synonyms:
超氧化钠
VWRC27996.230EA
1385
CNY
VWRC27996.230
VWRC27996.296
过氧化钠 ≥93.0%, powder, AnalaR NORMAPUR® analytical reagent
过氧化钠
分子式:
Na₂O₂ 分子量: 77.98 g/mol 溶点: 660 密度: 2.805 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00003497 CAS号: 1313-60-6 EINECS: 215-209-4 UN号: 1504 ADR: 5.1 I Merck Index: 13,08728 |
产品技术规格
Assay | Min. 93.0 % |
Heavy metals (as Pb) | Max. 20 ppm |
Total N (Nitrogen) | Max. 20 ppm |
Cl (Chloride) | Max. 40 ppm |
PO4 (Phosphate) | Max. 10 ppm |
SO4 (Sulphate) | Max. 50 ppm |
Al (Aluminium) | Max. 10 ppm |
Ca (Calcium) | Max. 0.05 % |
Fe (Iron) | Max. 20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...