氟化锂 99.98%, extra pure
供应商: Thermo Fisher Scientific
|
Danger
|
193380050
193380250
ACRO193380050EA
0
CNY
ACRO193380050
ACRO193380250
氟化锂 99.98%, extra pure
氟化锂
分子式:
FLi 分子量: 25.94 g/mol 沸点: 1676 溶点: 845 °C 密度: 2.63 g/cm³ 存储条件: Ambient |
MDL编号:
MFCD00011090 CAS号: 7789-24-4 EINECS: 232-152-0 UN号: 3288 ADR: 6.1 III Merck Index: 13,05553 |
产品技术规格
Appearance | White Powder |
Total trace metal impurities | ≤0.02 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...