铬酸钠, anhydrous 98+%
供应商: Thermo Fisher Scientific
|
Danger
|
377870250
377871000
ACRO377870250EA
0
CNY
ACRO377870250
ACRO377871000
铬酸钠, anhydrous 98+%
铬酸钠
分子式:
CrNa₂O₄ 溶点: 792 °C 密度: 2,698 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00003484 CAS号: 7775-11-3 EINECS: 231-889-5 UN号: 3290 ADR: 6.1,II Merck Index: 12,08745 |
产品技术规格
Appearance | Yellow Crystalline powder or crystals |
Titration Iodometric | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...