采血盘
供应商: VWR Collection
VWRI216-0816EA 915 CNY
VWRI216-0816 VWRI216-0814 VWRI216-0815 VWRI216-0817 VWRI216-0819
采血盘
盘子/皿/盆 血收集盘
ABS
  • Each tray comes with its own 13 or 16 mm test tube rack
  • Two disposable polystyrene inserts
  • Additional inserts for 13 or 16 mm test tube racks are available
Lightweight, roomy, and easy to clean blood collection tray. Keeping the tray clean and organised could not be easier. Along with the sturdy plastic frame, there are a set of economical, disposable inserts specifically designed for blood collection. Tray inserts come in two configurations, and include an extra-deep compartment for sharps. Other tray features include a supply drawer, sideMore Product Information