氯化铷 99.8+% (trace metals basis)
供应商: Thermo Fisher Scientific
Synonyms:
Rubidiumchloride
201260100
201260500
ACRO201260100EA
0
CNY
ACRO201260100
ACRO201260500
氯化铷 99.8+% (trace metals basis)
氯化铷
分子式:
RbCl 分子量: 120.92 g/mol 沸点: 1390 °C (760 mmHg) 溶点: 718 °C 存储条件: Ambient |
MDL编号:
MFCD00011187 CAS号: 7791-11-9 EINECS: 232-240-9 |
产品技术规格
Appearance | White Powder and/or chunks |
Total trace metal impurities | ≤2000.0 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...