Microcentrifuge tubes, 0,5/1,5/2,0 ml
供应商: BRAND
780757 780756 780758 780759 780760 780761 780762 780773 780774 780775 780755 780763 780764 780770 780771 780772
BRND780757EA 0 CNY
BRND780757 BRND780756 BRND780758 BRND780759 BRND780760 BRND780761 BRND780762 BRND780773 BRND780774 BRND780775 BRND780755 BRND780763 BRND780764 BRND780770 BRND780771 BRND780772
Microcentrifuge tubes, 0,5/1,5/2,0 ml
管子 微型离心管
PP, graduated, sterile or non sterile, with or without tamper-evident screw cap, autoclavable.
  • Can withstand centrifugation to 17000×g
  • DNA, DNase and RNase free, non-mutagenic, non-toxic
  • With frosted marking area
Operating range: −196 to +121 °C
Packaging: Pack of 500 sterile or 1000 non sterile version and caps.