甲基环戊烷 95%

供应商: Thermo Fisher Scientific
Danger

345245000 345240250
ACRO345245000EA 0 CNY
ACRO345245000 ACRO345240250
甲基环戊烷 95%
甲基环戊烷
分子式: C₆H₁₂
分子量: 84.16 g/mol
沸点: 71 °C
溶点: –142.4 °C
密度: 0.748 g/cm³
闪点: –4 °C
存储条件: Ambient
MDL编号: MFCD00001382
CAS号: 96-37-7
EINECS: 202-503-2
UN号: 2298
ADR: 3 II

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4110 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR