(溴甲基)环丁烷 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
环丁基甲基溴
367640050
367640250
ACRO367640050EA
0
CNY
ACRO367640050
ACRO367640250
(溴甲基)环丁烷 99%
(溴甲基)环丁烷
分子式:
C₅H₉Br 沸点: 145…148 °C (760 mmHg) 密度: 1,339 g/cm³ 闪点: 27 °C 存储条件: Ambient |
MDL编号:
MFCD00040960 CAS号: 17247-58-4 UN号: 1993 ADR: 3,III |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4790 to 1.4830 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...