4-Chlorophthalic anhydride ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 5-Chloroisobenzofuran-1,3-dione

C1328-25G C1328-5G
TCIAC1328-25GEA 0 CNY
TCIAC1328-25G TCIAC1328-5G
4-Chlorophthalic anhydride ≥98.0% (by GC, titration analysis)
4-Chlorophthalic anhydride
分子式: C₈H₃ClO₃
沸点: 290°C
MDL编号: MFCD00152354
CAS号: 118-45-6
EINECS: 204-251-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1483f3ce88b2f70d4559421d62de02a51a9cb4d0&vid=1483f3ce88b2f70d4559421d62de02a51a9cb4d0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 98.0 %
Melting point 95.0 to 100.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR