邻苯二甲酸二丙酯 99+%

供应商: Thermo Fisher Scientific
Warning

409160250 409161000
ACRO409160250EA 0 CNY
ACRO409160250 ACRO409161000
邻苯二甲酸二丙酯 99+%
邻苯二甲酸二丙酯
分子式: C₁₄H₁₈O₄
分子量: 250.29 g/mol
沸点: 317.5 °C (760 mmHg)
密度: 1.078 g/cm³
闪点: 166 °C
存储条件: Ambient
MDL编号: MFCD00009371
CAS号: 131-16-8
EINECS: 205-015-8
UN号: 3082

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4980 (20°C, 589 nm)
GC ≥99.0 %
Appearance of solution (0.5mmol/ml DMF) clear, colourless
Specific optical rotation +80.0° to +89.0° (20°C, 589 nm) (c=1, chloroform) <0.2% D-enantiomer
Residual solvents ≤5 % (2-Propanol, THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR