1-(2-氨乙基)哌嗪 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2-(1-哌嗪基)乙胺

400761000 400765000
ACRO400761000EA 0 CNY
ACRO400761000 ACRO400765000
1-(2-氨乙基)哌嗪 99%
1-(2-氨乙基)哌嗪
分子式: C₆H₁₅N₃
分子量: 129.21 g/mol
溶点: –19 °C
闪点: 88 °C
存储条件: Ambient
MDL编号: MFCD00005971
CAS号: 140-31-8
EINECS: 205-411-0
UN号: 2815
ADR: 8 III

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4990 to 1.5010 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR