Luperox AFR40, 过氧化苯甲酰 75% remainder water
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
过氧化苯甲酰,与邻苯二甲酸二丁酯的混合物
Stabilization:
Moistened with water
211780010
211780050
211781000
ACRO211780010EA
0
CNY
ACRO211780010
ACRO211780050
ACRO211781000
Luperox AFR40, 过氧化苯甲酰 75% remainder water
Luperox AFR40, 过氧化苯甲酰
分子式:
C₁₄H₁₀O₄ 分子量: 242.23 g/mol 溶点: 104…106 °C 存储条件: Ambient |
MDL编号:
MFCD00003071 CAS号: 94-36-0 EINECS: 202-327-6 UN号: 3104 ADR: 5.2 |
产品技术规格
Appearance | White Moist granular powder |
Titration Iodometric | 73 to 77 % |
Impurity | ≤0.25 % Inorganic and organic hydrolizable chloride |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...