1-(二苯基甲基)哌嗪 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 1-二苯甲基哌嗪

122931000 122935000
ACRO122931000EA 0 CNY
ACRO122931000 ACRO122935000
1-(二苯基甲基)哌嗪 97%
1-(二苯基甲基)哌嗪
分子式: C₁₇H₂₀N₂
溶点: 88…93 °C
闪点: 115 °C
存储条件: Ambient
MDL编号: MFCD00038379
CAS号: 841-77-0
EINECS: 212-667-7

现在订购

产品技术规格

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 88 to 93 °C
Water ≤3 % (K.F.)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR