5-氯-8-喹啉醇 95%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 5-氯-8-羟基喹啉

109301000 109300050
ACRO109301000EA 0 CNY
ACRO109301000 ACRO109300050
5-氯-8-喹啉醇 95%
5-氯-8-喹啉醇
分子式: C₉H₆ClNO
分子量: 179.61 g/mol
沸点: 349
溶点: 121…126 °C
存储条件: Ambient
MDL编号: MFCD00006788
CAS号: 130-16-5
EINECS: 204-978-1
Merck Index: 12,02483

现在订购

产品技术规格

Appearance Light green to grey Powder
Infrared spectrum Conforms
Melting point 121 to 126 °C
Titration with HClO4 ≥94 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR