2-氯喹啉 99%
供应商: Thermo Fisher Scientific
|
Warning
|
110040010
110040050
ACRO110040010EA
0
CNY
ACRO110040010
ACRO110040050
2-氯喹啉 99%
2-氯喹啉
分子式:
C₉H₆ClN 分子量: 163.61 g/mol 沸点: 265…267 °C (760 mmHg) 溶点: 34…37 °C 密度: 1.23 g/cm³ 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00006741 CAS号: 612-62-4 EINECS: 210-317-8 |
产品技术规格
Appearance | White to yellow Crystalline powder or crystals and/or chunks or fused mass |
Infrared spectrum | Conforms |
Melting point | 34 to 38 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...