黄尿酸 96%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4,8-二羟基喹啉甲酸, 4,8-二羟基喹啉-2-甲酸
115090010
115090050
ACRO115090010EA
0
CNY
ACRO115090010
ACRO115090050
黄尿酸 96%
黄尿酸
分子式:
C₁₀H₇NO₄ 分子量: 205.17 g/mol 溶点: 297…298 °C 存储条件: Ambient |
MDL编号:
MFCD00006754 CAS号: 59-00-7 EINECS: 200-410-1 Merck Index: 12,10202 |
产品技术规格
Appearance | Ochre Powder |
Infrared spectrum | Conforms |
Titration excess NaOH | ≥95.0 % |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...