喹啉-3-羧酸 98%

供应商: Thermo Fisher Scientific
Warning

175700050 175700010
ACRO175700050EA 0 CNY
ACRO175700050 ACRO175700010
喹啉-3-羧酸 98%
喹啉-3-羧酸
分子式: C₁₀H₇NO₂
分子量: 173.17 g/mol
沸点: 345
溶点: 277…283 °C
存储条件: Ambient
MDL编号: MFCD00006770
CAS号: 6480-68-8
EINECS: 229-337-3

现在订购

产品技术规格

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 277 to 283 °C
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR