Stoppers, standard, LaboPlast®
供应商: BÜRKLE
9763-1115 9763-1621 9763-2834 9763-3441 9763-4249 9763-0610 9763-0913 9763-1418 9763-1924 9763-2227 9763-2531 9763-3138 9763-4654
BURK9763-1115EA 17.63 CNY
BURK9763-1115 BURK9763-1621 BURK9763-2834 BURK9763-3441 BURK9763-4249 BURK9763-0610 BURK9763-0913 BURK9763-1418 BURK9763-1924 BURK9763-2227 BURK9763-2531 BURK9763-3138 BURK9763-4654
Stoppers, standard, LaboPlast®
塞子 聚乙烯/低密度聚乙烯/高密度聚乙烯塞
Laboratory standard bung made of PVC.
  • Good chemical resistance against acids and alkalis
  • Soft, red