3,5-二甲基苯酚 99+%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
5-羟基间二甲苯, 对称间二甲苯酚
162790050
162791000
162795000
ACRO162790050EA
0
CNY
ACRO162790050
ACRO162791000
ACRO162795000
3,5-二甲基苯酚 99+%
3,5-二甲基苯酚
分子式:
C₈H₁₀O 分子量: 122.17 g/mol 沸点: 222 °C (760 mmHg) 溶点: 63…66 °C 密度: 1.115 g/cm³ 闪点: 109 °C 存储条件: Ambient |
MDL编号:
MFCD00002307 CAS号: 108-68-9 EINECS: 203-606-5 UN号: 2928 ADR: 6.1 II Merck Index: 12,10215 |
产品技术规格
Appearance | White to yellow to orange Crystalline solid or fused mass |
Infrared spectrum | Conforms |
Melting point | 60 to 65 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...