3,4-二甲苯酚 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 3,4-二甲酚, 4-羟基邻二甲苯

116581000 116585000
ACRO116581000EA 0 CNY
ACRO116581000 ACRO116585000
3,4-二甲苯酚 99%
3,4-二甲苯酚
分子式: C₈H₁₀O
分子量: 122.17 g/mol
沸点: 227 °C (760 mmHg)
溶点: 63…67 °C
密度: 0.983 g/cm³
闪点: 80 °C
存储条件: Ambient
MDL编号: MFCD00002304
CAS号: 95-65-8
EINECS: 202-439-5
UN号: 2928
ADR: 6.1 II
Merck Index: 12,10215

现在订购

产品技术规格

Appearance White to yellow to brown Crystals or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 63 to 67 °C
GC ≥98.5 %
Color scale ≤250 APHA
Residue after ignition ≤0.005 %
Hydrocarbons Passes test
Chlorobenzene (C6H5Cl) ≤0.01 %
Nitrobenzene (C6H5NO2) (Limit about 0.001 %) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR