2,6-二异丙基苯胺 90+%
供应商: Thermo Fisher Scientific
169411000
169415000
ACRO169411000EA
0
CNY
ACRO169411000
ACRO169415000
2,6-二异丙基苯胺 90+%
2,6-二异丙基苯胺
分子式:
C₁₂H₁₉N 沸点: 257 °C (1013 hPa) 溶点: –45 °C 密度: 0,94 g/cm³ 闪点: 123 °C 存储条件: Ambient |
MDL编号:
MFCD00008887 CAS号: 24544-04-5 EINECS: 246-305-4 |
产品技术规格
Appearance | Colorless to yellow to orange to red to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5290 to 1.5370 (20°C, 589 nm) |
GC | ≥90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...