硫酸 95% in water ACS

供应商: Thermo Fisher Scientific
Danger

424520025 424525000
ACRO424520025EA 0 CNY
ACRO424520025 ACRO424525000
硫酸 95% in water ACS
硫酸
分子式: H₂SO₄
沸点: ∼335 °C (1013 hPa)
溶点: 10 °C
密度: 1,83 g/cm³
存储条件: Ambient
MDL编号: MFCD00064589
CAS号: 7664-93-9
EINECS: 231-639-5
UN号: 1830
ADR: 8,II
Merck Index: 14,08974

现在订购

产品技术规格

Appearance Free from suspended or insoluble matter
Color scale ≤10 APHA
Residue after ignition ≤0.0005 %
Titration with NaOH 95.0 to 98.0 %
Chloride (Cl) ≤0.2 ppm
Nitrate (NO3) ≤0.5 ppm
Ammonium (NH4) ≤2 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Mercury (Hg) ≤5 ppb
Heavy metals (as Pb) ≤1 ppm
KMnO4-reducing substance ≤2 ppm (as SO2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR