4-(2-氨乙基)-吗啉 98+%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2-氨乙基吗啉

400751000 400750050
ACRO400751000EA 0 CNY
ACRO400751000 ACRO400750050
4-(2-氨乙基)-吗啉 98+%
4-(2-氨乙基)-吗啉
分子式: C₆H₁₄N₂O
溶点: 24 °C
闪点: 175 °C
存储条件: Ambient
MDL编号: MFCD00006182
CAS号: 2038-03-1
EINECS: 218-011-6
UN号: 2923
ADR: 8,II

现在订购

产品技术规格

Appearance Clear colorless to white to yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4780 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR