(R)-(+)-1-(4-氯苯基)乙胺 97%, 98% ee

供应商: Thermo Fisher Scientific
Danger

370650010 370650050 370650250
ACRO370650010EA 0 CNY
ACRO370650010 ACRO370650050 ACRO370650250
(R)-(+)-1-(4-氯苯基)乙胺 97%, 98% ee
(R)-(+)-1-(4-氯苯基)乙胺
分子式: C₈H₁₀ClN
沸点: 232 °C (760 mmHg)
密度: 1,115 g/cm³
闪点: 115 °C
存储条件: Ambient
MDL编号: MFCD00671639
CAS号: 27298-99-3
UN号: 2810
ADR: 6.1,II

现在订购

产品技术规格

Appearance Clear colorless to pale yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5440 (20°C, 589 nm)
GC ≥96.0 %
Enantiomeric excess ≥97.5 %
Specific optical rotation +29° to +33° (20°C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR