2,4-二氯苯乙胺 98%

供应商: Thermo Fisher Scientific
Warning

350780050 350780250
ACRO350780050EA 0 CNY
ACRO350780050 ACRO350780250
2,4-二氯苯乙胺 98%
2,4-二氯苯乙胺
分子式: C₈H₉Cl₂N
分子量: 190.07 g/mol
密度: 1.26 g/cm³
闪点: >110 °C
存储条件: Ambient
MDL编号: MFCD00060611
CAS号: 52516-13-9

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5645 to 1.5665 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR