2-(3-氯苯基)乙胺 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 3-氯苯乙胺

342160050 342160250
ACRO342160050EA 0 CNY
ACRO342160050 ACRO342160250
2-(3-氯苯基)乙胺 98%
2-(3-氯苯基)乙胺
分子式: C₈H₁₀ClN
分子量: 155.63 g/mol
沸点: 111…113 °C (12 mmHg)
闪点: >109 °C
存储条件: Ambient
MDL编号: MFCD00047957
CAS号: 13078-79-0
EINECS: 235-982-1

现在订购

产品技术规格

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR