2-溴-6-甲氧基萘 98%
供应商: Thermo Fisher Scientific
|
Warning
|
190560050
190560250
ACRO190560050EA
0
CNY
ACRO190560050
ACRO190560250
2-溴-6-甲氧基萘 98%
2-溴-6-甲氧基萘
分子式:
C₁₁H₉BrO 分子量: 237.1 g/mol 溶点: 105…111 °C 存储条件: Ambient |
MDL编号:
MFCD00004062 CAS号: 5111-65-9 EINECS: 225-837-0 |
产品技术规格
Appearance | White to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 105 to 111 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...